missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79b Monoclonal Antibody (HM79-12), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen HMCD79B01
310.68 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00010kg
Description
CD79b Monoclonal antibody specifically detects CD79b in Mouse samples. It is validated for Flow Cytometry.Specifications
CD79b | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P15530 | |
Cd79b | |
Mouse CD79b. from CD79α/CD79β heterodimers purified from mouse (BALB/c x NZB)F1 B cell lymphoma cells, WEHI231 | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
HM79-12 | |
FITC | |
Cd79b | |
AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
Armenian Hamster | |
purified | |
RUO | |
15985 | |
4° C, store in dark | |
Liquid |