missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79a/mb-1 (B-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
135.66 EUR - 980.00 EUR
Specifications
Antigen | CD79a/mb-1 (B-Cell Marker) Ab-1 |
---|---|
Clone | HM47/A9 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12611167
|
Epredia
MS-357-B1 |
Concentrated | Biotin | 500 μL |
505.00 EUR
500µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12671157
|
Epredia
MS-357-B |
Concentrated | Biotin | 1 mL |
980.00 EUR
1mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12631167
|
Epredia
MS-357-P1 |
Concentrated | Unconjugated | 500 μL |
387.00 EUR
500µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12601167
|
Epredia
MS-357-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12641167
|
Epredia
MS-357-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12651167
|
Epredia
MS-357-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12691157
|
Epredia
MS-357-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12621167
|
Epredia
MS-357-P |
Concentrated | Unconjugated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12691167
|
Epredia
MS-357-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.Host Species: Mouse
Clone: HM47/A9
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Horse, Cow, Pig, Rabbit, Guinea Pig, Rat, Mouse and Chicken (Plasma cells). Does not react with opossum.
Epitope: GTYQDVGSLNIADVQ
Immunogen: A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein
Molecular Weight: 47 kDa
Positive Control: Raji cells, Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
CD79a/mb-1 (B-Cell Marker) Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
B-Cell | |
A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein | |
Primary |
HM47/A9 | |
Monoclonal | |
RUO | |
Bovine, Chicken, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
IgG2a κ |