missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 (Natural Killer Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments.
127.50 EUR - 640.00 EUR
Specifications
Antigen | CD57 (Natural Killer Cell Marker) Ab-1 |
---|---|
Clone | NK1 |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12639666
|
Epredia
MS-136-P |
Concentrated | 1 mL |
421.00 EUR
1mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12609676
|
Epredia
MS-136-R7 |
Prediluted | 7 mL |
345.00 EUR
7mL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12679666
|
Epredia
MS-136-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659666
|
Epredia
MS-136-P1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12649666
|
Epredia
MS-136-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12669666
|
Epredia
MS-136-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
CD57 is expressed on a subpopulation of 15-20% of peripheral blood mononuclear cells, about 60% of NK active cells and on a subset of T cells.Host Species: Mouse
Clone: NK1
Isotype: IgM/κ
Species Reactivity: Human. Does not react with rat. Others not known
Epitope: Not determined
Immunogen: Human peripheral blood mononuclear cells
Molecular Weight: 110 kDa
Positive Control: Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
1. Abo T Balch CM. J Immunol, 1982, 129(4):1758-61.
2. Abo T et al. J Immunology, 1982, 129:1752-7.
Specifications
CD57 (Natural Killer Cell Marker) Ab-1 | |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgM κ |
NK1 | |
Monoclonal | |
Mouse | |
Cell Adhesion | |
Human peripheral blood mononuclear cells | |
Primary |