missing translation for 'onlineSavingsMsg'
Learn More

CD56, APC, clone: MEM-188, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen A15704

Additional Details : Weight : 0.00280kg

 View more versions of this product

Product Code. 15285784

  • 228.00 EUR / 100 tests


Estimated Shipment date
from Supplier 01-02-2019
Add to basket

Specifications

Specifications

CD56, CD56 (NCAM1)
Monoclonal
APC
NCAM1
Liquid, Liquid
NCAM1
human CD56, human CD56
Purified
RUO
Antibody
Monoclonal
Human, Human
Flow Cytometry, Flow Cytometry
MEM-188, MEM-188
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P13591
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Mouse
IgG2a, kappa, IgG2a
100 tests
Store at 2 - 8 °C.
Primary
4684
Videos