Learn More
CD45RO Monoclonal Antibody (UCHL1), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD45RO05
Additional Details : Weight : 0.02000kg
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow Cytometry.Specifications
CD45RO | |
Monoclonal | |
APC | |
PTPRC | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
purified | |
ASR, ASR/IVD | |
5788 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
UCHL1 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P08575 | |
PTPRC | |
Human CD45RO | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |