missing translation for 'onlineSavingsMsg'
Learn More

CD45/T200/LCA Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, and flow cytometry experiments.

152.00 EUR - 428.00 EUR

Specifications

Antigen CD45/T200/LCA Ab-2
Clone PD7/26/16 + 2B11
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12611147
View Documents Promotion Details
Epredia
MS-355-P0
Concentrated 100 μL
152.00 EUR
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12641147
View Documents Promotion Details
Epredia
MS-355-P1
Concentrated 500 μL
298.00 EUR
0.50mL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12621147
View Documents Promotion Details
Epredia
MS-355-P
Concentrated 1 mL
388.00 EUR
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12681147
View Documents Promotion Details
Epredia
MS-355-R7
Prediluted 7 mL
359.00 EUR
7mL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12691147
View Documents Promotion Details
Epredia
MS-355-RQ
Ready-to-Use 12 mL
428.00 EUR
12mL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12631147
View Documents Promotion Details
Epredia
MS-355-P1ABX
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651147
View Documents Promotion Details
Epredia
MS-355-PABX
Concentrated 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

CD45 leucocyte common antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals. Antibody to CD45 is useful in differential identification of lymphoid tumors from non-hematopoietic undifferentiated neoplasms.

Host Species: Mouse

Clone: PD7/26/16 + 2B11

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat and dog. Others not known

Epitope: Not determined

Immunogen: PD7/26/16: human peripheral blood lymphocytes maintained in T-cell growth factor. 2B11: isolated neoplastic cells from T-cell lymphoma

Molecular Weight: 220, 205, 190, 180 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
Specifications

Specifications

CD45/T200/LCA Ab-2
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
PD7/26/16: human peripheral blood lymphocytes maintained in T cell growth factor. 2B11: isolated neoplastic cells from T cell lymphoma.
PD7/26/16 + 2B11
Monoclonal
Mouse
Oncogenesis
Primary
Videos
SDS
Documents

Documents

Product Certifications