missing translation for 'onlineSavingsMsg'
Learn More

CD45.2 Monoclonal Antibody (104), eFluor™ 450, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0454-80

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15391720

  • 102.00 EUR / 25µg
Estimated Shipment: 25-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The 104 monoclonal antibody reacts with the mouse CD45 molecule, the leukocyte common antigen (LCA) in CD45.2-expressing mouse strains. The strains that express CD45.2 include the most commonly used mouse strains C57BL/6, BALB/c, C58, DBA/1, DBA/2, C3H/He, CBA, 129, A and AKR. CD45.2 is expressed by all leukocytes in these strains. Applications Reported: This 104 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 104 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD45.2 Monoclonal antibody specifically detects CD45.2 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD45.2
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P06800
Ptprc
Affinity chromatography
RUO
19264
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
104
eFluor 450
Ptprc
B220; Cd45; CD45 antigen; CD45R; GP180; LCA; L-CA; leukocyte common antigen; loc; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type, C; Ptprc; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein
Mouse
25 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.