missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.1 Monoclonal Antibody (A20), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A14732
Additional Details : Weight : 0.01000kg
Description
CD45.1 Monoclonal antibody specifically detects CD45.1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD45.1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P06800 | |
Ptprc | |
Mouse CD45.1 from SJL mouse thymocytes and splenocytes | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
A20 | |
FITC | |
Ptprc | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Mouse | |
purified | |
RUO | |
19264 | |
4° C, store in dark | |
Liquid |