missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43/T-Cell Marker Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and flow cytometry applications with Epredia™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.
184.62 EUR - 399.00 EUR
Specifications
Antigen | CD43/T-Cell Marker Ab-1 |
---|---|
Clone | DF-T1 |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12689886
|
Epredia
MS-146-P1 |
Concentrated | 500 μL |
330.00 EUR
500µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12699886
|
Epredia
MS-146-P1ABX |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12609896
|
Epredia
MS-146-PABX |
Concentrated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12679886
|
Epredia
MS-146-P0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12639896
|
Epredia
MS-146-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytesHost Species: Mouse
Clone: DF-T1
Isotype: IgG1
Species Reactivity: Human. Does not react with rat. Others not tested
Epitope: Not determined
Immunogen: Myeloblastic KG1 cells
Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Specifications
CD43/T-Cell Marker Ab-1 | |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Myeloblastic KG1 cells |
DF-T1 | |
Monoclonal | |
Mouse | |
T-Cell | |
Primary |