missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Monoclonal Antibody (E11), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15455
Additional Details : Weight : 0.00280kg
Description
CD35 Monoclonal antibody specifically detects CD35 in Baboon, Chimpanzee, Cynomolgus Monkey, Human, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
CD35 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P17927 | |
CR1 | |
Human CD35 from Acute monocytic leukemia cells and normal blood monocytes | |
100 μg | |
Primary | |
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Rhesus Monkey | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
E11 | |
FITC | |
CR1 | |
C3 binding protein; C3b/C4b receptor; C3-binding protein; C3BR; C4BR; CD35; CD35 antigen; complement component (3b/4b) receptor 1 (Knops blood group); Complement Component (3b/4b) receptor 1 including Knops blood group system; complement component 3b/4b receptor 1 (Knops blood group); complement receptor type 1; CR1; KN; Knops blood group antigen | |
Mouse | |
purified | |
RUO | |
102117401, 1378, 449643, 718120 | |
4° C, store in dark | |
Liquid |