Learn More
CD33 Monoclonal Antibody (HIM3-4), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
148.00 EUR - 350.00 EUR
Specifications
Antigen | CD33 |
---|---|
Clone | HIM3-4 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15845493
|
Affymetrix eBioscience
62-0339-42 |
100 Tests |
350.00 EUR
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15835493
|
Invitrogen
62-0339-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The HIM3-4 monoclonal antibody reacts with human CD33, a 67 kDa member of the sialoadhesion family. CD33 is expressed by myelomonocytic precursors, monocytes, mast cells, and granulocytes. Hematopoietic stem cells and lymphocytes do not express this antigen. CD33 plays a role in sialic-acid dependent cell adhesion. Applications Reported: This HIM3-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIM3-4 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow Cytometry.Specifications
CD33 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P20138 | |
CD33 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD33 |
HIM3-4 | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
945 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |