missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Monoclonal Antibody (WM53), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15795
156.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.00280kg
Description
CD33 Monoclonal antibody specifically detects CD33 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD33 | |
Monoclonal | |
PE | |
CD33 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
945 | |
4° C | |
Liquid |
Flow Cytometry | |
WM53 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P20138 | |
CD33 | |
Human AML cells | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |