Learn More
CD33 Monoclonal Antibody (HIM3-4), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 62-0339-41
Additional Details : Weight : 0.01000kg
Description
Description: The HIM3-4 monoclonal antibody reacts with human CD33, a 67 kDa member of the sialoadhesion family. CD33 is expressed by myelomonocytic precursors, monocytes, mast cells, and granulocytes. Hematopoietic stem cells and lymphocytes do not express this antigen. CD33 plays a role in sialic-acid dependent cell adhesion. Applications Reported: This HIM3-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIM3-4 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser
Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.Specifications
CD33 | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P20138 | |
CD33 | |
Affinity chromatography | |
RUO | |
945 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HIM3-4 | |
Super Bright 436 | |
CD33 | |
CD33; p67; SIGLEC-3; SIGLEC3 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |