missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 Monoclonal Antibody (390), Alexa Fluor™ 647, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14716
Additional Details : Weight : 0.01000kg
Description
CD31 Monoclonal antibody specifically detects CD31 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD31 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
Q08481 | |
Pecam1 | |
Mouse leukocyte cell line 32D. | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
390 | |
Alexa Fluor 647 | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Rat | |
purified | |
RUO | |
18613 | |
4° C, store in dark | |
Liquid |