Learn More
CD31 Monoclonal Antibody (MBC78.2), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD3104
Additional Details : Weight : 0.50000kg
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for Flow Cytometry.Specifications
CD31 | |
Monoclonal | |
PE | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
purified | |
ASR, ASR/IVD | |
5175 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
MBC78.2 | |
PBS with 10% sucrose, 4mg/mL BSA and 0.1% sodium azide | |
P16284 | |
Pecam1 | |
Human CD31 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |