missing translation for 'onlineSavingsMsg'
Learn More

CD275 (B7-H2) Mouse anti-Human, Biotin, Clone: MIH12, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5889-82

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15258021

  • 189.00 EUR / 100µg


Estimated Shipment date
from Supplier 25-09-2019
Add to basket

Description

Description

Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
Specifications

Specifications

CD275 (B7-H2)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O75144
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, KIAA0653, LICOS, ICOS Ligand
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen)
MIH12
Biotin
ICOSLG
Liquid
ICOSLG
IgG1, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
23308
Videos
Documents

For Research Use Only.