missing translation for 'onlineSavingsMsg'
Learn More

CD22 / BL-CAM Mouse, Clone: BLCAM/1795, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 933-MSM5-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961198

  • 458.00 EUR / 100µg
Estimated Shipment: 03-10-2024
to see stock.

Description

Description

Specifications

Specifications

CD22 / BL-CAM
Cell Surface
BLCAM/1795
Unconjugated
NP_001172030
Mouse
130-140kDa
Protein A/G Purified
RUO
933
IgG1 κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
CD22
Recombinant human BL-CAM/CD22 protein (aa52-178)
Raji, Daudi, IM9, JY25 and human peripheral blood lymphocytes. Human Tonsil
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers