Learn More
CD184 (CXCR4) Monoclonal Antibody (12G5), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
361.00 EUR
Specifications
Antigen | CD184 (CXCR4) |
---|---|
Clone | 12G5 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15518716
|
Affymetrix eBioscience
25-9999-42 |
100 Tests |
361.00 EUR
100 tests |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 12G5 monoclonal antibody reacts with human CD184 (CXCR4), also termed Fusin, LESTR, or HUMSTR. CD184 is a member of the G-protein-coupled chemokine receptor family with seven membrane-spanning domains, and functions as a coreceptor for X4 HIV-1 entry into CD4+ cells. CD184 is expressed predomitly on naive T cell subsets of peripheral blood and is rapidly upregulated by PHA and IL-2 stimulation. The 12G5 antibody shows partial inhibition of chemotaxis and calcium influx induced by SDF-1 (the natural ligand of CD184), blocks CD4-independent HIV-2 infection, and blocks CD4-dependent infection by some T-tropic HIV-1 isolates. Applications Reported: This 12G5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 12G5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD184 (CXCR4) Monoclonal antibody specifically detects CD184 (CXCR4) in Human samples. It is validated for Flow Cytometry.Specifications
CD184 (CXCR4) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P61073 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
12G5 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
7852 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.