Learn More
CD144 (VE-cadherin) Monoclonal Antibody (16B1), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
175.00 EUR - 347.00 EUR
Specifications
Antigen | CD144 (VE-cadherin) |
---|---|
Clone | 16B1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15570197
|
Affymetrix eBioscience
53-1449-41 |
25 Tests |
175.00 EUR
25 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15580197
|
Affymetrix eBioscience
53-1449-42 |
100 Tests |
347.00 EUR
100 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling. Applications Reported: This 16B1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 16B1 antibody has been pre-titrated and tested by flow cytometric analysis of Human Umbilical Vein Endothelial Cells (HUVEC). This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD144 (VE-cadherin) Monoclonal antibody specifically detects CD144 (VE-cadherin) in Human samples. It is validated for Flow Cytometry.Specifications
CD144 (VE-cadherin) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
CDH5 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CDH5 |
16B1 | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
P33151 | |
1003 | |
IgG1 | |
Affinity chromatography | |
Antibody |
For Research Use Only.