missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11d Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568040
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
CD11d Polyclonal antibody specifically detects CD11d in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD11d | |
Polyclonal | |
Unconjugated | |
ITGAD | |
ADB2; beta-2 integrin alphaD subunit; CD11 antigen-like family member D; CD11d; FLJ39841; integrin alpha X (Cd11c); integrin alpha-D; integrin subunit alpha D; integrin, alpha D; ITGAD; Itgax; Leukointegrin alpha D | |
Rabbit | |
affinity chromatography | |
RUO | |
3681, 381924, 64350 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13349, Q3V0T4, Q9QYE7 | |
ITGAD | |
Synthesized peptide derived from human CD11d. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |