Learn More
Brady™ Caution Labels
Brand: Brady™ 225253
Additional Details : Weight : 0.05000kg
Specifications
10 Pk. | |
Black, Yellow on White | |
HORS SERVICE | |
4.33 in. | |
50 mm | |
0.249 mm | |
-40°F to +212°F | |
50 mm | |
Lockout Tagout Tags |
Tag | |
Polyester | |
Lockout Tagout | |
110 mm | |
2 in. | |
-40°C to +100°C | |
1.97 in. | |
7 mm |