missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CASP10 (Human) Recombinant Protein
Human CASP10 partial ORF ( NP_116756.2, 19 a.a. - 187 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 EUR - 508.00 EUR
Specifications
Accession Number | NP_116756.2 |
---|---|
Gene ID (Entrez) | 843 |
Name | caspase 10, apoptosis-related cysteine peptidase |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013725
|
Abnova™
H00000843-Q02.10UG |
10 μg |
335.00 EUR
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023725
|
Abnova™
H00000843-Q02.25UG |
25 μg |
508.00 EUR
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: SFREKLLIIDSNLGVQDVENLKFLCIGLVPNKKLEKSSSASDVFEHLLAEDLLSEEDPFFLAELLYIIRQKKLLQHLNCTKEEVERLLPTRQRVSLFRNLLYELSEGIDSENLKDMIFLLKDSLPKTEMTSLSFLAFLEKQGKIDEDNLTCLEDLCKTVVPKLLRNIEK
Specifications
NP_116756.2 | |
caspase 10, apoptosis-related cysteine peptidase | |
125% SDS-PAGE Stained with Coomassie Blue | |
ALPS2, FLICE2, MCH4 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
843 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CASP10 | |
GST |