Learn More
Erlab™ Captair Smart Filtering Fume Hood Mobicap
Erlab™ Captair Smart Filtering Fume Hood Mobicap offers a mobile, rolling bench for Captair Smart fume hoods.
Brand:  Erlab™ I0560000201
Additional Details : Weight : 30.00000kg
 View more versions of this product
Product Code. 15515184
  • 964.60 EUR / Each
Estimated Shipment: 28-08-2024
to see stock.
Description
Description
  • For Captair Smart filtering fume hoods
  • Equipped with 4 wheels and 2 self-locks
  • Allows the fume hood to be safely moved
Specifications
Specifications
Smart 321 Fume Hood
Without Work Surface
Metal
890 mm
Mobile Rolling Cart Work Bench