Learn More
Erlab™ Captair by Erlab™ Chemtrap V201 Version 2 Stand Alone Filtration System
ChemTrap™ is a stand alone filtration system for fireproof cabinets
Brand: Erlab™ G0201
Additional Details : Weight : 1.00000kg
Description
ChemTrap™ is a stand alone filtration system for fireproof cabinets. Units have been designed to vent and filter vapours coming from chemicals stored in fireproof safety cabinets. They allow fireproof safety cabinets to be totally autonomous by avoiding any connection to energy consuming air extraction systems.User protection
Eliminates noxious and odourous vapours generated by chemicals stored in safety cabinets. Optional filter saturation detector alerts users when it is necessary to replace filters.
Adaptability
Adaptable to any fireproof cabinet by connecting a multi diameter flexible pipe. These units are compatible with a large range of fireproof safety cabinets including Düperthal™ and Asecos™ cabinets.
Ambient air purification - no emissions
Working principle is based on the recirculation of filtered air. It contributes to purifying the air in the lab and also eliminates the direct emission of pollutants into the atmosphere.
Stand alone - energy saving
Quick and easy to install to any fireproof safety cabinet. It does not require an external air extraction system. Compared to a safety cabinet which is connected to a traditional energy consuming air extraction system, the very low energy consumption offers substantial savings on running costs. ChemTrap™ is available for vertical standing safety cabinets (H402 model) and for under bench safety cabinets (V201 model).
- Adaptable to a wide range of under bench and vertical safety cabinets
- Transforms safety cabinets into stand alone units
- Fast and easy installation
- Eliminates ducting to an extraction system
- Optional filter saturation detector
- Disconnect safety cabinets from fume hoods so that they can be switched off when not in use ChemTrap™ allows laboratories equipped with safety cabinets to benefit from protection against chemical inhalation risks.
Specifications
Stand Alone Filtration System | |
Fireproof Cabinets |