Learn More
Hirschmann™ Capillary Tubes, Both Ends Open
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ Capillary Tubes, Both Ends Open can be utilized for a wide range of scientific and medical applications.
Brand:  Hirschmann™ 9201312
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 10528201
  • 83.30 EUR / Pack of 1000
Estimated Shipment: 01-07-2024
Description
  • Both ends open
  • Available in a wide range of diameters/lengths
Specifications
Specifications
0.95 mm
Both ends open
Capillary Tube
1.35 mm
120 mm