Learn More
Applied Biosystems™ cAMP-Screen™ Cyclic AMP Immunoassay System
Ultrasensitive determination of cyclic AMP (cAMP) levels in cell lysates. Thermo Scientific cAMP-Screen™ 96-well Cyclic AMP immunoassay System is a competitive chemiluminescent immunoassay formatted with maximum flexibility to permit either manual r
Brand: Applied Biosystems™ 4412183
4585.63 EUR valid until 2024-12-31
Use promo code "21957" to get your promotional price.
Additional Details : Weight : 0.00450kg
Description
- Utilizes highly sensitive chemiluminescent CSPD™ Substrate with Sapphire-II™ Enhancer that is triggered by an enzyme conjugate composed of cAMP bound to alkaline phosphatase (cAMP-AP)
- Ready-to-use chemiluminescent substrate/enhancer formulation generates sustained-glow light emission from 30 minutes after addition
- Mainly used in secondary screening and pre-clinical research, where sensitivity and no false positives are essential
- Highest sensitivity of any commercially available cAMP assay
- Hours of read time with little or no degradation of signal
- Simple protocol
- Useful for wide range of receptor activation studies
This chemiluminescent assay is designed to provide the highest sensitivity of any commercially available cAMP assay. As few as 60 femtomoles of cAMP can be detected. The assay has a wide dynamic range, detecting from 0.06 to 6,000 picomoles of cAMP without the need for sample dilution or manipulations such as acetylation. This is especially helpful in cell-based assays when measuring Gs- or Gi-coupled agonist or antagonist stimulation and/or inhibition. Intra-assay precision for duplicate samples is typically 5% or less. Once the substrate/enhancer formulation reaches glow signal, the plate can be read for hours with little or no degradation of the signal. This is useful in screens where several plates are compared with each other. In addition, the assay exhibits exceptionally low cross-reactivity with other adenosine-containing or cyclic nucleotides.
- Target: Cyclic AMP
- Assay Incubation Time: 1 hr.
- Assay Sensitivity: 60 femtomoles cAMP
- Detection Method: Chemiluminescent
- For use with Equipment: Microplate Reader
- Label/Dye: Alkaline Phosphatase
- Sample Type: Cell Lysates
- Sample Volume: 60μL
- Storage: Store at 2° to 8°C
Specifications
60 femtomoles cAMP | |
Chemiluminescence | |
GPCR (G-Protein Coupled Receptors) | |
Microplate Reader | |
Non-Gene | |
960 assays | |
Cyclic AMP |
1 kit, store at 2°C to 8 °C | |
cAMP-Screen™ | |
Cell Lysates | |
Non-Gene | |
AP | |
60 μL |
For Research Use Only. Not for use in diagnostic procedures.