Learn More
Invitrogen™ CAMK2G (CaMKII gamma) Recombinant Human Protein
CAMK2G (CaMKII gamma) is a ubiquitous serine/threonine protein kinase that has been implicated in diverse effects of hormones and neurotransmitters. Recombinant human CAMK2G (c-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.
1678.00 EUR - 7490.00 EUR
Specifications
For Use With (Application) | Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation |
---|---|
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol |
Molecular Weight (g/mol) | 60.9kDa |
Storage Requirements | Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
Protein Length | aa c-terminus. Contains S36P SNP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478923
|
Invitrogen™
PV6270 |
1mg |
7490.00 EUR
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15468923
|
Invitrogen™
PV6269 |
100μg |
1678.00 EUR
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CAMK2G has six alternatively spliced variants that encode six different isoforms. Some of these variants have been identified in human tumors. Transgenic mice expressing a partially calcium-independent mutant form of CAMKG showed 1.5- to 2-fold increase in the thymus of these mice, at least in part due to an increase in the life span of double-positive thymocytes. There was an increase in the number of T cells in the secondary lymphoid organs that had acquired an antigen-dependent memory phenotype.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
60.9kDa | |
aa c-terminus. Contains S36P SNP | |
Recombinant |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Human | |
GST Tag |