Learn More
Alfa Aesar™ Calpain Substrate I
For cellular and molecular biology applications
Brand:  Alfa Aesar™ J63222.MC
Additional Details : CAS Number : 94367-20-1 Weight : 0.00010kg
 View more versions of this product
Product Code. 15485479
  • 277.00 EUR / 100mg
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Specifications
White
Lyophilized
Fluorogenic
Flourogenic substrate for both calpains I and II
551.59
100mg
Videos
SDS
For Research Use Only