missing translation for 'onlineSavingsMsg'
Learn More

Calibration Buffer pH 7.01, FDA Bottle, Hanna Instruments™

Catalog Number 11577989
Click to view available options
Quantity:
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
Boiling Point Boiling Point: ∽100°C
Color Colorless
pH 7.01
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Formulation Aqueous Buffer Solution
Odor Odorless
Packaging Bottle
Traceability to NIST Yes
Solubility Information Soluble in water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'