missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate, ACS Reagent, ≥99%, Honeywell Fluka™

Catalog Number 15605300
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
10124-36-4
CdO4S
208.47
MFCD00010923
QCUOBSQYDGUHHT-UHFFFAOYSA-L
cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
24962
CHEBI:50292
cadmium(2+);sulfate
[O-]S(=O)(=O)[O-].[Cd+2]
This item is not returnable. View return policy
10124-36-4
CdO4S
208.47
MFCD00010923
QCUOBSQYDGUHHT-UHFFFAOYSA-L
cadmium sulfate, cadmium sulphate, cadmium monosulfate, sulfuric acid, cadmium salt 1:1, cadmium sulfuricum, cadmium sulfate 1:1, cadmium sulfate, tetrahydrate, cadmium 2+ sulfate, caswell no. 136c, unii-947unf3z6o
24962
CHEBI:50292
cadmium(2+);sulfate
[O-]S(=O)(=O)[O-].[Cd+2]

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 1000°C
Color Colorless
Quantity 100 g
Linear Formula CdSO4
UN Number UN2570
Formula Weight 208.47g/mol
Percent Purity ≥99%
Loss on Drying ≤1.0%
Packaging Poly Bottle
Appearance Crystalline
Density 4.69g/cm3
Chemical Name or Material Cadmium sulfate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'