Learn More
Fisherbrand™ Bunsen Burner Neoprene Tubing
Suitable for use with coal and natural (methane) gas burners.
6.80 EUR - 66.25 EUR
Product Code | Brand | Overall Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Overall Length (Metric) | Price | Quantity & Availability | |||||
12330768
|
Fisherbrand™
|
10 m |
66.25 EUR
10m |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11855522
|
Fisherbrand™
|
600 mm |
8.45 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11845522
|
Fisherbrand™
|
450 mm |
6.80 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11865522
|
Fisherbrand™
|
750 mm |
9.95 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11875522
|
Fisherbrand™
|
900 mm |
9.75 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10492624
|
Fisherbrand™
|
2 m |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
With reinforced ends for fitting to Bunsen burner and gas tap.Specifications
Neoprene | |
Bunsen Burner |