Learn More
Buffer Solution (Biphthalate), pH 4 (Color Coded Red), BAKER ANALYZED Reagent, J.T. Baker™
Brand:  JT Baker 5657-01
Additional Details : CAS Number : 50-00-0 Weight : 0.60000kg
Product Code. 15597614
  • 87.05 EUR / 500mL
Estimated Shipment: 03-07-2024
Specifications
Buffer Solution (Biphthalate)
500 mL
Poly container