Learn More
Buffer solution, 11.0, boric acid / sodium hydroxide solution / potassium chloride, Honeywell Fluka™
Catalog Number p-7072081
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 100 mL
6 x 500 mL
6 x 1 L
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
N.O
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
CHEBI:18219
azanium;hydroxide
N.O
Boric acid / sodium hydroxide solution / potassium chloride
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
pH 11.0
Quantity 100 mL
UN Number NONH for all modes of transport
Chemical Name or Material Buffer solution