Learn More
Fisherbrand™ Buffer Recirculation Port
Accessory for Maxi-Cooled Horizontal Gel Electrophoresis Unit.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
Product Code. 12315498
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Used for reinforcement of ions
  • May be connected to peristaltic pump for buffer recirculation during electrophoresis
  • Maintains buffer pH
  • Prevents ionic gradient formation
Specifications
Specifications
Horizontal Unit Maxi Gel Electrophoresis Unit
Buffer Recirculation Port
Two
1/Pk.