Learn More
Fisherbrand™ Buffer Recirculation Port
Accessory for Maxi-Cooled Horizontal Gel Electrophoresis Unit.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Used for reinforcement of ions
- May be connected to peristaltic pump for buffer recirculation during electrophoresis
- Maintains buffer pH
- Prevents ionic gradient formation
Specifications
Horizontal Unit Maxi Gel Electrophoresis Unit | |
Buffer Recirculation Port | |
Two | |
1/Pk. |