Learn More
Invitrogen™ eBioscience™ BrdU Kit for IHC/ICC Colorimetric
Contains the necessary reagents and buffers for identifying and examining proliferating cells by immunohistochemical or immunocytochemical analysis. Optimized for IHC with both frozen and paraffin-embedded BrdU-labeled mouse intestine and ICC of BrdU-pulsed HeLa cells grown on culture slides.
Brand:  Invitrogen™ 8800-6599-45
Additional Details : Weight : 0.23750kg
Product Code. 15511247
  • 497.00 EUR / 50 tests
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Alert:
Contains Sodium azide, formaldehyde, and hydrogen peroxide
Includes: BrdU Fixation Buffer
BrdU Antigen Retrieval Solution (10X)
BrdU Blocking Solution
Anti-BrdU Biotin (clone BU20A) : 10 μg
Avidin-HRP
3, 3'-Diaminobenzidine (DAB) Substrate Solution: Reagent A (DAB Chromogen), Reagent B (DAB Diluent), and Dropper Bottle (for storage and application of working DAB solution)
Description
Description
  • Cells are labeled in vitro or in vivo with 5-bromo-2′-deoxyuridine (BrdU), which is incorporated into DNA in place of thymidine during the S-phase of the cell cycle
  • Following fixation and antigen-retrieval steps, cells or tissue sections are stained for BrdU incorporation and visualized using an Avidin-HRP and DAB substrate reaction
Specifications
Specifications
BrdU
DAB
eBioscience™
IHC/ICC BrdU Kit
Colorimetric
HRP (Horseradish Peroxidase) Substrate
ICC (Immunochemistry), IHC (Immunohistochemistry)
All species
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.