Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Oil Mist Filters
Oil Mist Filters and replacements for VACUUBRAND Rotary Vane Vacuum Pumps
Brand:  VACUUBRAND™
374.75 EUR valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10485823
  • 451.00 EUR / Each
Estimated Shipment: 24-07-2024
Description
  • For use with VACUUBRAND Rotary Vane Vacuum Pumps
  • Oil Mist Filters “FO” trap up to 99% of oil mist generated by the pump, helping preserve the laboratory environment
Specifications
Specifications
Model RE8, RE9, RE16 and RZ8, RZ9, RZ16 rotary vane vacuum pump
Oil Mist Filter