missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAF [V599E] Recombinant Human Protein, Invitrogen™
BRAF acts upstream of MEK in the MAP kinase pathway.
1826.00 EUR - 7890.00 EUR
Specifications
Accession Number | NP_004324.1, V599E |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 67.1kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458553
|
Invitrogen
PR7304A |
100μg |
1826.00 EUR
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10210783
|
Invitrogen
PV4173 |
1mg |
7890.00 EUR
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
BRAF is involved in cell division, differentiation, and secretion. The BRAF mutant V599E is oncogenic and found in a variety of cancers.
Specifications
NP_004324.1, V599E | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
RAF Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
67.1kDa | |
RUO - research use only | |
BRAF V599E | |
Human | |
GST Tag | |
Frozen |