Learn More
Brady™ BradyJet™ J2000 Polypropylene Labels
Chemical resistant, low temperature labels ideal for full-color identification and barcode applications
Brand: Brady™ 150542
Additional Details : Weight : 0.15100kg
Description
Specifications
Acrylic | |
Matte | |
BradyJet™ J2000 | |
100 ft x 1.125 in. | |
100 ft. x 1.125 in. | |
1.5 in. |
White | |
Polypropylene | |
1 Ea. | |
0.005 in. | |
100 ft. |