Learn More
Brady™ BradyJet™ J2000 Polypropylene Labels
Chemical resistant, low temperature labels ideal for full-color identification and barcode applications
Brand: Brady™ 150543
Additional Details : Weight : 0.50000kg
Description
Specifications
Acrylic | |
Matte | |
BradyJet™ J2000 | |
3.35 x 2.17 in. | |
2.50 in. |
White | |
Polypropylene | |
340/Pk. | |
0.005 in. | |
3.35 in. |