Learn More
Trajan™ BPX5 GC Capillary Columns
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
Brand:  Trajan™ 054113
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10268810
  • 780.00 EUR / Each
Estimated Shipment: 11-06-2024
Description
  • Very low bleed
  • High temperature
  • Non-polar
  • Ideal for GCMS
  • Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Specifications
0.22 mm
370°C
0.25 μm
25 m