Learn More
RETSCH Bottom Cones
Brand:  RETSCH 42.787.0011
19380.00 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 11964742
  • 20400.00 EUR / Each
Estimated Shipment: 27-05-2024
Specifications
Bottom Cone
110 mm
For PT 200