Learn More
Fisherbrand™ Stirrer Bosshead
Designed to fit 18mm diameter vertical rods and 13mm horizontal support rods.
Brand:  Fisherbrand™
Additional Details : Weight : 0.20000kg
Product Code. 12946826
  • 44.60 EUR / Each
Estimated Shipment: 25-07-2024
Description
  • Constructed in anodized aluminum with stainless steel socket screws
  • Supplied with allen key and adapters to convert 18mm hole to accept 13mm rod and the 13mm hole to accept a 9.5mm rod
  • Blue finish
Specifications
Specifications
Anodized Aluminum, Stainless Steel
Allen key, adapters
18mm vertical rods, 13mm horizontal support rods