missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified

Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish

Brand:  Karl Hecht™ 42110018

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10083871

  • 18.97 EUR / Each
Estimated Shipment: 31-01-2024
to see stock.
Add to basket

Description

Description

  • Duran™ borosilicate glass 3.3.
  • Standard type.
  • Standard ground and interchangeable NS stopper of polyethylene.
  • Fused blue graduation.
  • Thick-walled, solid finish.
  • Clearly legible figures and marks.
  • Conformity certified.
    • Individual test- or lot-control certificate available from manufacturer with surcharge.
Specifications

Specifications

Polyethylene
Borosilicate Glass
0.025 mL
NS Stopper
5 mL
Conical
Conical
NS 7/16
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers