Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Marke: Lenz Laborglasinstrumente™ 04030049
Weitere Details : Weight : 0.10000kg
Beschreibung
- Cylindrical shape with straight discharge tube
- With NS socket and PE stopper
- With solid glass plug
- With Screwthreade retaining device
- With scale
Spezifikation
250 mL | |
Borosilicate Glass | |
Polyethylene |