Learn More
Wheaton™ Borosilicate Glass Mortar
Brand: Wheaton™ 358010
162.45 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.45360kg
Specifications
Yes | |
Borosilicate Glass | |
For Tissue Grinder |
15mL | |
Mortar |