Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Filter Funnels
Lenz Laborglasinstrumente™ Filter funnels are ideal for separating solids from liquids via laboratory filtration.
Brand:  Lenz Laborglasinstrumente™ 07195134
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10652092
  • 45.79 EUR / Each
Estimated Shipment: 25-06-2024
Description
  • Made from borosilicate glass
  • Separates solids from liquids
Specifications
Specifications
75 mL
130 mm
1
Borosilicate Glass
45mm
Filter funnel