Learn More
Robu™ Borosilicate Glass Crude Filter Crucibles
Marke: Robu™ 20604
Weitere Details : Weight : 0.10000kg
Spezifikation
34mm | |
For crude fiber extraction | |
34mm | |
Crude Filter Crucible |
Borosilicate Glass | |
30 mL | |
60mm | |
4 |