Learn More
Robu™ Borosilicate Glass Buchner Filter Funnels
Brand:  Robu™ 21404
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 10104411
  • 510.00 EUR / Each
Estimated Shipment: 26-06-2024
Specifications
190mm (OD); 180mm (ID)
30mm
206 mm
190mm
Buchner filter funnel
4000 mL
Borosilicate Glass
Highly resistant to most chemicals such as solvents, acids and alkalis
4