Learn More
Thermo Scientific Chemicals Bombesin
For cellular and molecular biology applications
Brand: Thermo Scientific Chemicals J63514.#0
Additional Details : CAS Number : 31362-50-2 Weight : 0.00005kg
Description
Gut tetradecapeptide with the ability to stimulate release of various hormones
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Powder | |
1619.90 | |
5 mg | |
Glp-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-His-Leu-Met-NH2 |
2°C to 8°C | |
Gut tetradecapeptide with the ability to stimulate release of various hormones | |
Bombesin | |
RUO - research use only |
RUO – Research Use Only