Learn More
Bohlender™ BOLA™ Spare Washer for Stopcocks
Spare washer for stopcocks made of polypropylene or PTFE.
Brand:  Bohlender™ J1198-21
Additional Details : Weight : 0.01000kg
Product Code. 17137474
  • 58.88 EUR / Pack of 20
Estimated Shipment: 05-06-2024
Description
  • Washers made of PP or PTFE
  • For BOLA™ Eco Stopcock Plugs
  • Color: Red
Specifications
Specifications
Thread M 8
Polypropylene
For BOLA™ Eco Stopcock Plugs
Taper 1:10/1:10/1:5
NS 12.5, NS 14.5, 13.5